Learn More
Selleck Chemical LLC Ditiocarb sodium E7036-1G
SDP
Supplier:  Selleck Chemical LLC E70361G
Encompass_Preferred
Ditiocarb sodium (Sodium diethyldithiocarbamate) is an accelerator of the rate of copper cementation Sodium diethyldithiocarbamate reduces the incidence of HIV infection and also enhances adjuvant immunoresearch of high risk breast cancer
Catalog No. 50-000-40924