Learn More
Medchemexpress LLC HY-14218 1g Medchemexpress, Ditolylguanidine CAS:97-39-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY142181G
Encompass_Preferred
Medchemexpress, HY-14218 1g Ditolylguanidine CAS:97-39-2 Ditolylguanidine (1,3-Di-o-tolylguanidine) is an agonist of sigma receptor (σ1/σ2 receptor) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2699