Learn More
Medchemexpress LLC Diuron-d6 | 1007536-67-5 | MFCD01632291 | 99.0% | 239.13 | C9H4D6Cl2N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0860S10MG
Encompass_Preferred
Diuron-d6 is the deuterium-labeled form of the herbicide diuron provided as a stable-isotope analytical standard for laboratory research and quantitative analysis. It is intended for use as an internal standard in mass spectrometry and for environmental and biological residue studies where isotopic labeling improves accuracy and detection.
Catalog No. 50-002-98197