Learn More
Selleck Chemical LLC Divalproex Sodium S1703-50mg
SDP
Supplier:  Selleck Chemical LLC S170350MG
Encompass_Preferred
Divalproex Sodium consisting of a compound of sodium valproate and valproic acid in a 1 1 molar relationship in an enteric coated form is a HDAC inhibitor used in the treatment for epilepsy
Catalog No. 50-311-0092