Learn More
Enterprise Technology Solutions Diversey Care AirFreshener,Liquid,Concentrated,GreenApple,2.52.5L,2/CT,Clear
SDP
Supplier:  Enterprise Technology Solutions DVO101109733
Encompass_Preferred
Good Sense HC Concentrated Liquid Air Freshener uses fast-acting odor counteractants to neutralize and eliminate odors instead of masking them. Effective, Fresh-scented concentrate effectively neutralizes odors from tobacco, food, mold, mildew, urine and vomit. Formula is safe for carpets, hard surfaces and in the air. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.
Catalog No. 50-007-90480