Learn More
Enterprise Technology Solutions Diversey Care Cleaner,Neutral pH,Citrus,Multipurpose,2.5 Liter,2/CT,Orange
SDP
Supplier:  Enterprise Technology Solutions DVO904716
Encompass_Preferred
Neutral cleaner features a nonalkaline formula that dries film-free to prevent dulling floor finishes. Easy-to-use solution does not require rinsing for quick, simple cleaning. Low-foaming design allows less emptying of recovery tanks and increased performance. Concentrated cleaner with a citrus scent is Green Seal and Kosher certified. Neutral cleaner is perfect for everyday cleanings of nearly all washable surfaces.
Catalog No. 50-008-30678