Learn More
Enterprise Technology Solutions Diversey Care Cleaner Spray,Heavy-duty,f/Carpet,Conc,1 Gal,4/CT,Clear
SDP
Supplier:  Enterprise Technology Solutions DVO904266
Encompass_Preferred
Heavy Duty Prespray cleans heavily soiled carpets in heavy-traffic areas. Carpets stay cleaner longer thanks to exclusive soil barriers. These barriers made with a unique polymer-enhanced formula help resist resoiling. Concentrated carpet cleaner has a 1:12 dilution ratio and a fruity scent.
Catalog No. 50-008-30458