Learn More
Enterprise Technology Solutions Diversey Care Extraction Rinse,Carpet,NoResidue,FruitScent,84.5oz,2/CT,CR
SDP
Supplier:  Enterprise Technology Solutions DVO101109761
Encompass_Preferred
Carpet Extraction Rinse effectively removes hard water deposits and neutralizes residue from previous cleanings. Versatile formula is strong enough for advanced-generation carpet fibers at low dilution, yet delicate enough for wool carpets at high dilution. Specialty rinse also makes salt residue more soluble, meaning less passes to remove winter salt, leaving to tacky residue.
Catalog No. 50-007-90545