Learn More
Enterprise Technology Solutions Diversey Care Maintainer/Rejuvenator, Revive Plus, 2.5 Liter, 2/CT, RD
SDP
Supplier:  Enterprise Technology Solutions DVO92974618
Encompass_Preferred
Revive Plus SC Maintainer/Rejuvenator features a unique formula that repairs microscopic scratches to maintain a glossy look for your floors. Gentle ingredients allow safe use through daily cleanings without browning out. Advanced solution enhances your floor's appearance when burnished with 1500 to 2000 plus rpm propane, battery or electric burnishers. Cleaner/maintainer is designed for use as a hard-surface maintainer.
Catalog No. 50-008-30993