Learn More
Medchemexpress LLC DiZPK hydrochloride | 2349295-23-2 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12801A10MM/1ML
Encompass_Preferred
DiZPK hydrochloride is a structural analog of pyrrolysine (Pyl) that functions as a photocrosslinker. It is used for identifying direct protein-protein interactions in living prokaryotic and eukaryotic cells. This product is intended for research use only.
  • Structural analog of pyrrolysine (Pyl)
  • Photocrosslinker for identifying direct protein-protein interactions
  • Used in living prokaryotic and eukaryotic cells
  • For research use only
Catalog No. 50-003-39747