Learn More
Medchemexpress LLC HY-133146 5mg Medchemexpress, DJ001 CAS:2161305-12-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1331465MG
Encompass_Preferred
Medchemexpress, HY-133146 5mg DJ001 CAS:2161305-12-8 DJ001 is a highly specific, selective and non-competitive protein tyrosine phosphatase-σ (PTPσ) inhibitor with an IC50 of 1.43 μM. DJ001 displays no inhibitory activity against other phosphatases, with only modest inhibitory activity against Protein Phosphatase 5. DJ001 promotes promote hematopoietic stem cell regeneration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9851