Learn More
Selleck Chemical LLC Skatole S4959-1g
SDP
Supplier:  Selleck Chemical LLC S49591G
Encompass_Preferred
Skatole (3-methylindole Scatole) is a mildly toxic white crystalline organic compound that occurs naturally in feces It has a fairly broad bacteriostatic effect
Catalog No. 50-311-2122