Learn More
Medchemexpress LLC Djenkolic acid | 498-59-9 | MFCD00063079 | 98.0% | 254.33 g·mol^-1 | C7H14N2O4S2 | 25MG

Supplier: Medchemexpress LLC HY12592325MG
Djenkolic acid analytical reference standard intended for research and analytical applications. This sulfur-containing non-protein amino acid (CAS 498-59-9) is provided as a small-scale reference material for assay development, method validation, and analytical quality control. Refer to the product SDS and Certificate of Analysis for handling and storage instructions.
- Intended for use as an analytical reference standard.
- CAS number 498-59-9.
- Molecular formula C7H14N2O4S2; molecular weight 254.33 g·mol⁻¹.
- Reported purity 98.0%.
- Pack size 25 mg.
- Handling and storage per SDS and certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.