Learn More
Medchemexpress LLC Djenkolic acid | 498-59-9 | MFCD00063079 | 98.0% | 254.33 g/mol | C7H14N2O4S2 | 100MG

Supplier: Medchemexpress LLC HY125923100MG
Djenkolic acid analytical standard (CAS 498-59-9) is a sulfur-containing non-protein amino acid supplied as a reference material for analytical and methodological research. It is intended for use in chromatography and mass spectrometry workflows for qualitative and quantitative assays. Typical pack sizes range from 5 mg to 500 mg, with the 100 mg pack commonly used for laboratory calibration and method validation.
- Analytical reference standard for HPLC, GC, and MS.
- Purity 98.0%.
- Molecular weight 254.33 g/mol.
- Molecular formula C7H14N2O4S2.
- Available in multiple pack sizes, including 100 mg.
- Suitable for method development and quantitative analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.