Learn More
Medchemexpress LLC DK1 | 1187568-17-7 | 99.7% | C16H13ClN2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14322650MG
Encompass_Preferred
DK1 is a potent modulator of estrogen related receptor. It has the ability to reduce blood glucose and impacts the activity of the ERRα receptor. DK1 has potential for diabetes research.
  • Potent modulator of estrogen related receptor.
  • Ability to reduce blood glucose.
  • Impacts the activity of ERRα receptor.
  • Potential for the research of diabetes.
Catalog No. 50-003-55515