Learn More
Medchemexpress LLC DKFZ-748 | 2490709-68-5 | C18H23N3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1515905MG
Encompass_Preferred
DKFZ-748 is a selective HDAC10 inhibitor that demonstrates anti-tumor activity. This compound shows dose-dependent growth inhibition of HeLa cells in a polyamine-limiting in vitro tumor model. It also induces HDAC acetylation of BE(2)-C cells and leads to significant accumulation of N8 acetyl- and N1,8-diacetylspermidine.
  • Selective HDAC10 inhibitor
  • Exhibits anti-tumor activity
  • Induces dose-dependent growth inhibition in HeLa cells
  • Promotes HDAC acetylation of BE(2)-C cells
  • Leads to significant accumulation of N8 acetyl- and N1,8-diacetylspermidine
Catalog No. 50-003-72938