Learn More
Selleck Chemical LLC DKM 2-93
SDP
Supplier:  Selleck Chemical LLC S584125MG
Encompass_Preferred
DKM 2-93 is a relatively selective lead inhibitor of ubiquitin-like modifier activating enzyme 5 (UBA5)
Catalog No. 50-000-03900