Learn More
Medchemexpress LLC DL-2-amino-3-phosphonopropionic acid | 5652-28-8 | MFCD00014350 | 98.0% | 169.07 g/mol | C3H8NO5P | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10078650MG
Encompass_Preferred
DL-AP3 (DL-2-amino-3-phosphonopropionic acid) is a research-grade small molecule that functions as a competitive antagonist of metabotropic glutamate receptors mGluR1 and mGluR5 and as an inhibitor of phosphoserine phosphatase. It has reported neuroprotective effects and is supplied for laboratory research use only.
  • Acts as a competitive antagonist of mGluR1 and mGluR5.
  • Inhibits phosphoserine phosphatase activity.
  • Reported to exhibit neuroprotective effects in experimental models.
  • High purity for research applications (98.0%).
  • Molecular weight 169.07 g/mol; molecular formula C3H8NO5P.
  • Provided as a solid suitable for biochemical and pharmacological assays.
Catalog No. 50-002-13403