Learn More
Medchemexpress LLC DL-2-amino-5-phosphonovaleric acid | 76326-31-3 | MFCD00010515 | 98.0% | 197.13 | C5H12NO5P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100714100MG
Encompass_Preferred
DL-AP5 (DL-2-amino-5-phosphonovaleric acid) is a competitive NMDA receptor antagonist used as a research reagent to probe glutamatergic signaling. It is supplied as a white to off-white solid, with reported high purity and standard storage recommendations for powder and solutions, and is used in both in vitro and in vivo studies to investigate NMDA-mediated responses and related physiological effects.
  • Competitive NMDA receptor antagonist for research use.
  • High reported purity suitable for biochemical assays.
  • White to off-white solid, convenient for handling and formulation.
  • Available in small research pack sizes for laboratory experiments.
  • Stable as a powder under frozen storage for long-term use.
  • Solutions have defined short-term storage stability for experimental workflows.
  • Used in vitro and in vivo to study NMDA-mediated neuronal responses.
Catalog No. 50-002-14361