Learn More
Selleck Chemical LLC DL-2-Aminoadipic acid S5442-1g
SDP
Supplier:  Selleck Chemical LLC S54421G
Encompass_Preferred
DL-2-Aminoadipic acid is an excitatory amino acid analogue which functions as a glutamine synthetase inhibitor
Catalog No. 50-311-0339