Learn More
Medchemexpress LLC DL-2-aminooctanoic acid | 644-90-6 | MFCD00008102 | 98.0% | 159.23 g/mol | C8H17NO2 | 5 G

Supplier: Medchemexpress LLC HYW0154615G
2-Aminooctanoic acid (DL-α-aminocaprylic acid) is an alpha-amino fatty acid supplied as the racemic DL mixture. It is used as a synthetic building block and research reagent in peptide synthesis, organic synthesis, and biochemical studies where an aminated caprylic acid derivative is required.
- Racemic DL mixture for research applications.
- Purity 98.0%.
- Molecular formula C8H17NO2; molecular weight 159.23 g/mol.
- Powder form suitable for coupling and synthetic transformations.
- Available in multiple small pack sizes for lab use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.