Learn More
Medchemexpress LLC DL-3-phenyllactic acid | 828-01-3 | MFCD00065928 | >98.0% | 166.18 | C9H10O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW017162R
Encompass_Preferred
DL-3-phenyllactic acid (CAS 828-01-3) is an analytical standard and endogenous metabolite used in research and analytical applications. It is a solid organic acid (C9H10O3, MW 166.18) commonly used as a reference compound in biochemical, microbiological, and metabolomics studies.
  • Provides a reference standard for analytical assays.
  • Suitable for metabolite identification and quantification.
  • Solid form simplifies weighing and storage.
  • Compatible with common analytical techniques such as HPLC and GC-MS.
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-003-03951