Learn More
Medchemexpress LLC DL-5-hydroxylysine hydrochloride | 13204-98-3 | MFCD00012878 | 97.3% | 198.65 g/mol | C6H15ClN2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW014930R50MG
Encompass_Preferred
DL-5-Hydroxylysine hydrochloride is the racemic (DL) hydrochloride salt of 5-hydroxylysine supplied as a research-grade reference standard for analytical and biochemical applications, including studies of collagen post-translational modification and amino acid analysis.
  • Racemic (DL) hydrochloride salt of 5-hydroxylysine.
  • Used as a reference standard for analytical and biochemical assays.
  • High purity, approximately 97.3% as reported on the certificate of analysis.
  • White to off-white solid; supplied in small vials for laboratory use.
  • Molecular weight 198.65 g/mol; formula C6H15ClN2O3.
  • Store refrigerated (≈4 °C) per manufacturer recommendations.
Catalog No. 50-003-02515