Learn More
Medchemexpress LLC DL-5-hydroxylysine hydrochloride | 13204-98-3 | MFCD00012878 | 99.9% | 198.65 g·mol⁻¹ | C6H15ClN2O3 | 50 MG

Supplier: Medchemexpress LLC HYW01493050MG
DL-5-hydroxylysine hydrochloride is the hydrochloride salt of DL-5-hydroxylysine, a post-translationally modified, non-essential amino acid primarily found in collagen. It is supplied as a solid reagent for biochemical and metabolic research.
- Hydrochloride salt form, water-soluble.
- High purity: 99.85%.
- White to off-white solid appearance.
- Molecular weight 198.65 g·mol⁻¹; formula C6H15ClN2O3.
- Solubility in water ≥100 mg/mL (≈503.4 mM).
- CAS number 13204-98-3.
- Store sealed, away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.