Learn More
Cayman Chemical DL-a-Difluoromethylornithine
SDP
Supplier:  Cayman Chemical 16889100 MG
Encompass_Preferred
An irreversible inhibitor of ornithine decarboxylase that suppresses polyamine biosynthesis; displays antiangiogenic, cytostatic, and antiparasitic effects
Catalog No. 50-296-6094