Learn More
Medchemexpress LLC DL-A-phosphatidylcholine, dipalmitoyl-13C40 | 15397-46-3 | 13C40H80NO8P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY150707S5MG
Encompass_Preferred
DL-A-phosphatidylcholine, dipalmitoyl-13C40 is the 13C labeled DL-A-phosphatidylcholine, dipalmitoyl. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Isotope-labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-003-75645