Learn More
Medchemexpress LLC DL-Alanine-13C-3 | 131157-42-1 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN2362S1
Encompass_Preferred
DL-Alanine-13C-3 is a 13C-labeled DL-Alanine, an amino acid, and the racemic compound of L- and D-alanine. This laboratory chemical is intended for research use only by experienced personnel in appropriately equipped and authorized facilities. It plays a key role in the glucose-alanine cycle between tissues and the liver.
  • Used as a reducing and capping agent for the production of nanoparticles
  • Suitable for research on transition metal chelation, including Cu(II), Zn(II), and Cd(II)
  • Classed as a sweetener, alongside glycine and sodium saccharin
Catalog No. 50-004-21907