Learn More
Selleck Chemical LLC DL-Alanine E4832-1G
SDP
Supplier:  Selleck Chemical LLC E48321G
Encompass_Preferred
DL-alanine (DL-2-Aminopropionic acid) is a racemic mixture of L- and D-alanine that forms complexes with transition metals like Cu(II) Zn(II) and Cd(II) It also plays a key role in the glucose-alanine cycle between tissues and the liver
Catalog No. 50-000-43316