Learn More
Medchemexpress LLC DL-Allylglycine | 7685-44-1 | MFCD00063103 | 98.0% | 115.13 g/mol | C5H9NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW023493
Encompass_Preferred
DL-Allylglycine (2-aminopent-4-enoic acid) is a research-grade amino acid derivative that acts as a glutamate decarboxylase (GAD) inhibitor. It is used in neuroscience studies to reduce GABA levels and to produce convulsant activity for seizure and epilepsy models. The compound has formula C5H9NO2 and molecular weight 115.13 g/mol.
  • Glutamate decarboxylase (GAD) inhibitor used in neuroscience research.
  • Used to induce convulsant activity for seizure and epilepsy models.
  • Chemical formula C5H9NO2 and molecular weight 115.13 g/mol.
  • High purity suitable for analytical and in vivo studies.
  • Available in multiple pack sizes, including 25 g.
  • Documentation includes data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-003-03681