Learn More
Apexbio Technology LLC DL-AP3 5652-28-8 100mg
SDP
Supplier:  Apexbio Technology LLC A4535100
Encompass_Preferred
DL-AP3 (CAS 5652-28-8) is a small molecule that acts as a competitive antagonist of group I metabotropic glutamate receptors (mGluR1 and mGluR5) thereby modulating glutamatergic neurotransmission Additionally DL-AP3 inhibits phosphoserine phosphatase affecting serine metabolism Through these actions DL-AP3 is commonly used in research to investigate the physiological and pathological functions of group I mGluRs in neural signaling as well as to study pathways involving serine biosynthesis and metabolism
Catalog No. 50-000-96205