Learn More
Selleck Chemical LLC DL-AP5 E2979-25MG
SDP
Supplier:  Selleck Chemical LLC E297925MG
Encompass_Preferred
DL-AP5 (2-APV DL-2-Amino-5-phosphonovaleric acid) is a competitive antagonist of NMDA (N-methyl-D-aspartate) receptor It shows significantly antinociceptive activity and specifically blocks on channels in the rabbit retina
Catalog No. 50-000-40520