Learn More
Medchemexpress LLC DL-Arginine-13C6,15N4 hydrochloride | 95.0% | 220.59 g·mol⁻1 | 13C6H15Cl15N4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW014375S410MG
Encompass_Preferred
DL-Arginine-13C6,15N4 hydrochloride is an isotopically labeled racemic arginine hydrochloride used as an internal standard, tracer, or labeled amino acid in mass spectrometry, metabolic flux analysis, and SILAC workflows. It produces predictable mass shifts for quantitative proteomics and metabolic studies, and is supplied as a solid powder with supporting documentation.
  • Isotopically labeled with 13C6 and 15N4 for clear mass shifts.
  • Suitable as an internal standard for quantitative mass spectrometry.
  • Racemic (DL) arginine hydrochloride form, supplied as a solid powder.
  • Available in small mg-scale packages for tracer and labeling experiments.
  • Includes product datasheet and certificate of analysis for traceability.
Catalog No. 50-003-02862