Learn More
Medchemexpress LLC DL-arginine-13C6,15N4 hydrochloride | 95.0% | 220.59 g/mol | 13C6H15Cl15N4O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW014375S41MG
Encompass_Preferred
DL-Arginine-13C6,15N4 hydrochloride is an isotopically labeled DL-arginine supplied as the hydrochloride salt for research use. It is intended for mass spectrometry, stable isotope tracing, and metabolic studies, and is provided in small research-scale quantities with manufacturer documentation.
  • Isotopically labeled with 13C6 and 15N4 for mass spectrometry and tracer studies.
  • Supplied as hydrochloride salt to improve solubility and handling.
  • High purity (95%) suitable for analytical applications.
  • Available in small milligram pack sizes for method development and labeling experiments.
  • Datasheet, certificate of analysis, and safety data sheet available for quality and safety information.
Catalog No. 50-003-02125