Learn More
Medchemexpress LLC DL-Arginine-13C6 hydrochloride | 99.0% | 216.62 g/mol | 13C6H15ClN4O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW014375S21MG
Encompass_Preferred
DL-Arginine-13C6 hydrochloride is a 13C6-isotopically labeled, racemic arginine supplied as the hydrochloride salt for use as an internal standard, tracer, or labeled reagent in mass spectrometry and metabolic studies. It is intended for analytical applications and should be stored sealed and protected from moisture for optimal stability.
  • High isotopic enrichment (13C6) for clear mass shifts in MS analyses.
  • High purity: 99.0% by HPLC, suitable for analytical work.
  • Hydrochloride salt form provides good solubility in aqueous media.
  • Stable under recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Small supplied quantity tailored for tracer and standard preparation.
Catalog No. 50-003-03301