Learn More
Medchemexpress LLC Dl-arginine-13C6 hydrochloride | 99.0% | 216.62 g/mol | 13C6H15ClN4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW014375S25MG
Encompass_Preferred
DL-Arginine-13C6 hydrochloride is the 13C6-labeled racemic form of arginine supplied as the hydrochloride salt. It is intended for use as an internal standard, tracer, or labeled substrate in biochemical and mass-spectrometry studies, supporting accurate quantitation and metabolic tracing.
  • 13C6 isotopic labeling for tracer and quantitative studies.
  • Hydrochloride salt form for improved stability and solubility.
  • Racemic DL composition suitable for general labeling applications.
  • Small research pack sizes for method development and standard curves.
  • Molecular weight approximately 216.62 g/mol for reference in calculations.
Catalog No. 50-003-02402