Learn More
Medchemexpress LLC DL-aspartic acid-13C,15N-1 hydrochloride | 98.0% | 171.55 | C4H8ClNO4 | 100 UG

Supplier: Medchemexpress LLC HYW015824S5100UG
DL-Aspartic acid-13C,15N-1 hydrochloride is the 13C- and 15N-labeled form of DL-aspartic acid used as an isotopic tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as the hydrochloride salt with high purity for analytical workflows.
- Isotope-labeled with 13C and 15N for unambiguous mass-spectral and NMR identification.
- Suitable as a tracer or internal standard for NMR, GC-MS, and LC-MS quantitation.
- High purity (98.0%) supporting reliable analytical results.
- Molecular weight 171.55 g/mol for calculating concentrations and conversions.
- Provided as a hydrochloride salt for improved stability and handling.
- Available in small-scale quantities appropriate for analytical and tracer studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.