Learn More
Medchemexpress LLC DL-Buthionine-S R-su 50mg | 50MG
SDP
Supplier:  Medchemexpress LLC HY106376B50MG
Encompass_Preferred
DL-Buthionine-(S R)-sulfoximine hydrochloride (Buthionine sulfoximine hydrochloride) is a potent inhibitor of glutamylcysteine synthetase biosynthesis
Catalog No. 50-002-42258