Learn More
Medchemexpress LLC DL-Buthionine-(S,R)-sulfoximine | 5072-26-4 | MFCD00070309 | >98.0% | 222.31 g/mol | C8H18N2O3S | 250 MG
SDP
Supplier:  Medchemexpress LLC HY106376250MG
Encompass_Preferred
DL-Buthionine-(S,R)-sulfoximine is a small-molecule research reagent that irreversibly inhibits glutamylcysteine synthetase, producing rapid depletion of cellular glutathione. It is used in cell and animal models to study oxidative stress, ferroptosis, and glutathione-dependent pathways. Not for human or clinical use.
  • Irreversible inhibitor of glutamylcysteine synthetase.
  • Depletes cellular glutathione to model oxidative stress.
  • Suitable for studies of ferroptosis and redox biology.
  • Provided as a characterized, laboratory-grade chemical for research use.
  • Supports reproducible biochemical and cell-based assays.
Catalog No. 50-002-43772