Learn More
Medchemexpress LLC Dl-buthionine-(S,R)-sulfoximine (hydrochloride) | 00-00-0 | MFCD00070309 | 99.1% | 258.77 | C8H19ClN2O3S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY106376B500MG
Encompass_Preferred
DL-Buthionine-(S,R)-sulfoximine hydrochloride is the hydrochloride salt form of buthionine sulfoximine, a cell-permeable, potent, and irreversible inhibitor of γ-glutamylcysteine synthetase. It is commonly used to deplete cellular glutathione for studies of redox biology, oxidative stress, and ferroptosis, and is supplied as a powder for preparation of solutions for in vitro and in vivo experiments.
  • Inhibits γ-glutamylcysteine synthetase, reducing glutathione synthesis.
  • Cell-permeable and fast-acting for cellular assays.
  • Useful for redox biology, oxidative stress, and ferroptosis research.
  • Provided as a hydrochloride salt powder for solution preparation.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-43471