Learn More
Medchemexpress LLC DL-Cystathionine | 535-34-2 | MFCD00136759 | 97.38% | 222.26 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW009749A10MG
Encompass_Preferred
DL-Cystathionine is an isomeric mixture of four forms of cystathionine and allocystathionine. It serves as an intermediate compound in the biological pathway responsible for the synthesis of the amino acid cysteine. This chemical is designated for research purposes only and is not intended for human or therapeutic use.
  • Isomeric mixture of cystathionine and allocystathionine.
  • Intermediate in amino acid cysteine biosynthesis.
  • Intended solely for research applications.
  • Available in solid form, white to off-white in color.
  • Purity of 97.38%.
Catalog No. 50-004-53384