Learn More
Medchemexpress LLC DL-Cysteine-d1 | 3374-22-9 | 95.0% | 122.16 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW012850S110MG
Encompass_Preferred
DL-Cysteine-d1 is the deuterated labeled 2-Amino-3-mercaptopropanoic acid, which is a cysteine derivative. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium have the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Amino acids and their derivatives can serve as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage.
Catalog No. 50-004-55049