Learn More
Cayman Chemical DL-CystIn-d4 50mg
SDP
Supplier:  Cayman Chemical 3090250 MG
Encompass_Preferred
An internal standard for the quantification of cystine by GC- or LC-MS
Catalog No. 50-298-8434