Learn More
Medchemexpress LLC DL-cystine-d6 | 352431-53-9 | MFCD01317433 | 99.8% | 246.34 g·mol⁻¹ | C6H6D6N2O4S2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW013940S11MG
Encompass_Preferred
DL-Cystine-d6 is the deuterium-labeled form of DL-cystine provided for research use as an isotopic tracer and analytical standard. It is typically used in mass spectrometry, metabolic tracing, and quantitative labeling studies where deuterium incorporation allows clear differentiation from unlabeled compounds.
  • Deuterium-labeled tracer for isotope studies
  • High purity (99.81%) suitable for analytical applications
  • Molecular formula C6H6D6N2O4S2, molecular weight 246.34 g·mol⁻¹
  • Available in small pack sizes for analytical workflows
  • Solid form, stable under recommended storage conditions
Catalog No. 50-003-03936