Learn More
Cayman Chemical DL-erythrothreo SphngnIn 10mg
SDP
Supplier:  Cayman Chemical 2437610 MG
Encompass_Preferred
A mixture of sphinganine (d16:0), L-erythro sphinganine (d16:0), D-threo sphinganine (d16:0), and L-threo sphinganine (d16:0); decreased in rats following long-term, low-dose administration of dimethoate and is used as a biomarker for dimethoate exposure; found in the fermentation broth of Bacterium SRCnm
Catalog No. 50-301-6031