Learn More
TARGETMOL CHEMICALS INC DL-Goitrin 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2S177825MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DL-Goitrin is a potent antithyroid compound found naturally in crucifers. DL-Goitrin is responsible for the induction of glutathione S-transferases. DL-Goitrin is a moderate inhibitor of purified bovine adrenal dopamine beta-hydroxylase, leads to a depression of brain norepinephrine and to an elevation of heart and adrenal dopamine. Purity 99.94%
Catalog No. 50-287-9275