Learn More
Cayman Chemical DL-HIstIdIn-d3 5mg
SDP
Supplier:  Cayman Chemical 348415 MG
Encompass_Preferred
An internal standard for the quantification of histidine by GC- or LC-MS
Catalog No. 50-291-1295