Learn More
Medchemexpress LLC DL-Homocysteine-d4 | 416845-90-4 | 98.4% | 139.21 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW010347S1MG
Encompass_Preferred
DL-Homocysteine-d4 is the deuterium labeled DL-Homocysteine. DL-Homocysteine is a weak neurotoxin, and can affect the production of kynurenic acid in the brain. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66466