Learn More
Medchemexpress LLC DL-Isocitric acid trisodium salt | 1637-73-6 | 98.0% | 258.07 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW009362500MG
Encompass_Preferred
DL-Isocitric acid trisodium salt is an endogenous metabolite and an intermediate product in the citric acid cycle. It can be used as a marker for determining the composition of isocitrates in fruit products. It can also inhibit the hypertrophy of cardiomyocytes in vitro.
  • Endogenous metabolite
  • Intermediate product in the citric acid cycle
  • Can be used as a marker for determining the composition of isocitrates in fruit products
  • Can inhibit the hypertrophy of cardiomyocytes in vitro
Catalog No. 50-003-66989