Learn More
Medchemexpress LLC DL-lanthionine | 3183-08-2 | MFCD00002615 | 98.0% | 208.24 | C6H12N2O4S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW3278081MG
Encompass_Preferred
DL-Lanthionine is the DL stereoisomer of lanthionine, a sulfur-containing, non-proteinogenic amino acid used as a research reagent and synthetic building block in studies of peptide antibiotics and sulfur-dependent peptide chemistry. Supplied as a powder for laboratory use.
  • Used as a building block in peptide antibiotic research.
  • Monosulfide counterpart to cystine, useful for sulfur chemistry studies.
  • High purity: 98.0%.
  • Available in small research pack sizes, including 1 mg.
  • Powder storage stability: -20°C for up to 3 years.
  • Chemical formula C6H12N2O4S; molecular weight 208.24.
Catalog No. 50-002-80690