Learn More
Medchemexpress LLC DL-laudanosine | 1699-51-0 | 99.5% | 1 ML

Supplier: Medchemexpress LLC HY12248910MM 1ML
DL-Laudanosine is a metabolite of the neuromuscular blocking agents atracurium and cisatracurium that crosses the blood-brain barrier and may cause central nervous system excitation and seizure activity. The product is available as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid material in multiple pack sizes for research use.
- Ready-to-use 10 mM solution in DMSO, 1 mL vial.
- High purity suitable for research and analytical applications.
- Molecular formula C21H27NO4 and molecular weight 357.44 g/mol.
- Purity reported at 99.53%.
- Useful as a reference metabolite for pharmacological studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.